Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Supplier: Thermo Fisher Scientific
|
Warning
|
42496.AD
42496.AK
42496.K2
42496.ADEA
60.5
EUR
42496.AD
42496.AK
42496.K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Polydimethylsiloxane, trimethylsiloxy terminated
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Flash Point: 39°(102°F)
Physical Form: Liquid
Density: 0.820
Refractive Index: 1.3825
Appearance: Colourless
Odour: Odourless
Melting Point: -86°
Boiling Point: 152-153°
UN No.: UN1993
MDL No.: MFCD00148360
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: 152…153 °C Melting Pt: –86 °C Density: 0.820 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 UN: 1993 ADR: 3,III |