Methyl purple 0,1% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Synonyms: Fleisher's Indicator Solution
42753.AP 42753.K2
42753.APEA 52.7 EUR
42753.AP 42753.K2
Methyl purple 0,1% (w/v) in aqueous solution
Methyl purple
Physical Form: Liquid MDL No.: MFCD00070843
Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
MW: 1391.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070843
CAS Number: 1340-02-9