Fullerene nanotube multi-walled 20 nm OD, Length: 5-20 micron
Supplier: Thermo Fisher Scientific
|
Warning
|
43839.MD
43839.MF
43839.MDEA
198
EUR
43839.MD
43839.MF
Fullerene nanotube multi-walled 20 nm OD, Length: 5-20 micron
Fullerene nanotube
CAS No.: 308068-56-6
Physical Form: -270 mesh powder
Appearance: Black
UN No.: UN1325
MDL No.: MFCD00133992
Storage Temperature:
Ambient |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 UN: 1325 ADR: 4.1,III |