Fullerene nanotube multi-walled 20-50 nm OD, Length: 5-20 micron
Supplier: Thermo Fisher Scientific
|
Warning
|
44192.03
44192.06
44192.03EA
276
EUR
44192.03
44192.06
Fullerene nanotube multi-walled 20-50 nm OD, Length: 5-20 micron
Fullerene nanotube
CAS No.: 308068-56-6
Physical Form: Powder
Appearance: Black
UN No.: UN1325
MDL No.: MFCD00133992
Storage Temperature:
Ambient |
MDL Number:
MFCD00133992 CAS Number: 308068-56-6 UN: 1325 ADR: 4.1,III |