5-Vinyluracil ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Vinylpyrimidine-2,4(1H,3H)-dione
44379.01
44379.03
44379.01EA
119
EUR
44379.01
44379.03
5-Vinyluracil ≥97%
5-Vinyluracil
Purity: 97%
CAS No.: 37107-81-6
Molecular Formula: C6H6N2O2
Formula Weight: 138.12
Physical Form: Powder
MDL No.: MFCD00913267
Formula:
C₆H₆N₂O₂ MW: 138.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00913267 CAS Number: 37107-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...