2-Chlorotrityl chloride 1,0 - 1,4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Fisher Scientific
Synonyms: (2-Chlorophenyl)diphenylmethyl chloride, Chloro(2-chlorophenyl)diphenylmethane, (Chloro(2-chlorophenyl)methylene)dibenzene
44407.04 44407.09 44407.18
44407.04EA 115 EUR
44407.04 44407.09 44407.18
2-Chlorotrityl chloride 1,0 - 1,4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh
2-Chlorotrityl chloride
CAS No.: 42074-68-0 Physical Form: Powder MDL No.: MFCD00040399
Formula: C₁₉H₁₄Cl₂
MW: 313.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040399
CAS Number: 42074-68-0