Platinum 70% (nominal) on high surface area advanced carbon support, HiSPEC® 13100
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Platinium
45041.06 45041.03
45041.06EA 0 EUR
45041.06 45041.03
Platinum 70% (nominal) on high surface area advanced carbon support, HiSPEC® 13100
Platinum
CAS No.: 7440-06-4 Appearance: Dark grey to black Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 1325
ADR: 4.1 II
Merck Index: 12,07689