Tetraammineplatinum(II) hydroxide hydrate (≥58% Pt)
Supplier: Thermo Fisher Scientific
Warning
46319.03 46319.06
46319.03EA 267 EUR
46319.03 46319.06
Tetraammineplatinum(II) hydroxide hydrate (≥58% Pt)
Tetraammineplatinum(II) hydroxide hydrate
CAS No.: 312695-70-8 Molecular Formula: H14N4O2Pt.xH2O Formula Weight: 297.23(anhy) Physical Form: Powder Appearance: Off-white MDL No.: MFCD00151253
Formula: H₁₄N₄O₂Pt·xH₂O
MW: 297.23 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151253
CAS Number: 312695-70-8
EINECS: 253-823-4