n-Heptane, anhydrous, molecular sieves_over
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
high purity solvents
Argon_Chemseal
47191.AK
47191.K2
47191.AKEA
50
EUR
47191.AK
47191.K2
n-Heptane, anhydrous, molecular sieves_over
n-Heptane
CAS No.: 142-82-5
Molecular Formula: C7H16
Formula Weight: 100.21
Flash Point: -4°(24°F)
Physical Form: Liquid
Density: 0.684
Refractive Index: 1.3877
Appearance: Colourless
Odour: Petroleum-like
Melting Point: -91°
Boiling Point: 97-99°
Beilstein Reference: 1730763
UN No.: UN1206
MDL No.: MFCD00009544
Formula:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Boiling Pt: 97…99 °C Melting Pt: –91 °C Density: 0.684 Flash Pt: –4 °C (24 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3 II Merck Index: 14,4659 |