6-Chlorothiochroman-4-one 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A10304.03
A10304.06
A10304.03EA
27.8
EUR
A10304.03
A10304.06
6-Chlorothiochroman-4-one 98%
6-Chlorothiochroman-4-one
Purity: 98%
CAS No.: 13735-12-1
Molecular Formula: C9H7ClOS
Formula Weight: 198.67
Melting Point: 65-69°
Beilstein Reference: 124621
MDL No.: MFCD00052997
Formula:
C₉H₇ClOS Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052997 CAS Number: 13735-12-1 |