Dowex® 1X2 (Cl) 50-100 mesh
Supplier: Thermo Fisher Scientific
A12130.0B
A12130.18
A12130.30
A12130.0BEA
913
EUR
A12130.0B
A12130.18
A12130.30
Dowex® 1X2 (Cl) 50-100 mesh
Dowex® 1X2 (Cl)
CAS No.: 9085-42-1
Odour: Amine-like
MDL No.: MFCD00132715
Storage Temperature:
Ambient |
MDL Number:
MFCD00132715 CAS Number: 9085-42-1 |