1,3-Divinyltetramethyldisiloxane 96% cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane
Supplier: Thermo Fisher Scientific
|
Warning
|
A12463.14
A12463.22
A12463.14EA
58.4
EUR
A12463.14
A12463.22
1,3-Divinyltetramethyldisiloxane 96% cont. up to 4% 1-vinyl-3-ethyltetramethyldisiloxane
1,3-Divinyltetramethyldisiloxane
Purity: 96%
CAS No.: 2627-95-4
Molecular Formula: C8H18OSi2
Formula Weight: 186.40
Flash Point: 24°(75°F)
Refractive Index: 1.4110
Melting Point: -100°
Boiling Point: 139-140°
Beilstein Reference: 1762585
UN No.: UN1993
MDL No.: MFCD00014933
Formula:
C₈H₁₈OSi₂ Boiling Pt: 139…140 °C Melting Pt: –100 °C Density: 0.811 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014933 CAS Number: 2627-95-4 EINECS: 220-099-6 UN: 1993 ADR: 3,III |