Perfluorophenanthrene mixture of isomers
Supplier: Thermo Fisher Scientific
Synonyms: Perfluoroperhydrophenanthrene
A18898.14 A18898.22
A18898.14EA 79 EUR
A18898.14 A18898.22
Perfluorophenanthrene mixture of isomers
Perfluorophenanthrene
CAS No.: 306-91-2 Molecular Formula: C14F24 Formula Weight: 624.12 Refractive Index: 1.3348 Melting Point: -20° Boiling Point: 215° MDL No.: MFCD00042596
Formula: C₁₄F₂₄
MW: 624.116 g/mol
Boiling Pt: 215 °C
Melting Pt: –20 °C
Density: 2.030
Storage Temperature: Ambient
MDL Number: MFCD00042596
CAS Number: 306-91-2
EINECS: 400-470-0