2-Ethylacrolein 90% stabilised, tech.

Supplier: Thermo Fisher Scientific
Danger


Stabilization: Stabilised with hydroquinone (HQ) 50 ppm

B21839.06 B21839.14 B21839.22
B21839.06EA 61.1 EUR
B21839.06 B21839.14 B21839.22
2-Ethylacrolein 90% stabilised, tech.
2-Ethylacrolein

Stab. with 50ppm hydroquinone


Purity: 90% CAS No.: 922-63-4 Molecular Formula: C5H8O Formula Weight: 84.12 Storage Temperature: 2°C to 8°C Flash Point: 1°(34°F) Refractive Index: 1.4245 Odour: Pungent Boiling Point: 92-93° Beilstein Reference: 1098378 UN No.: UN1992 MDL No.: MFCD00010129

Formula: C₅H₈O
Boiling Pt: 92…93 °C
Density: 0.859
Flash Pt: 1 °C (34 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00010129
CAS Number: 922-63-4
EINECS: 213-079-3
UN: 1992
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR