Benzyl ethyl malonate ≥85%, tech.

Supplier: Thermo Fisher Scientific

B23011.14 B23011.22
B23011.14EA 134 EUR
B23011.14 B23011.22
Benzyl ethyl malonate ≥85%, tech.
Benzyl ethyl malonate

Purity: 85% CAS No.: 42998-51-6 Molecular Formula: C12H14O4 Formula Weight: 222.24 Refractive Index: 1.4970 Boiling Point: 145°/4mm MDL No.: MFCD00009194

Formula: C₁₂H₁₄O₄
Boiling Pt: 145 °C (4 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00009194
CAS Number: 42998-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR