3,5-Bis(trifluoromethyl)styrene 98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
B23499.03
B23499.06
B23499.03EA
82.7
EUR
B23499.03
B23499.06
3,5-Bis(trifluoromethyl)styrene 98% stabilised
3,5-Bis(trifluoromethyl)styrene
Purity: 98%
CAS No.: 349-59-7
Molecular Formula: C10H6F6
Formula Weight: 240.15
Storage Temperature: 2°C to 8°C
Flash Point: 50°(122°F)
Refractive Index: 1.4250
Melting Point: 7-8°
Boiling Point: 59-61°/20mm
UN No.: UN1993
MDL No.: MFCD00075538
Formula:
C₁₀H₆F₆ MW: 240.15 g/mol Boiling Pt: 59…61 °C (20 mmHg) Melting Pt: 7…8 °C Density: 1.334 Flash Pt: 50 °C (122 °F) |
MDL Number:
MFCD00075538 CAS Number: 349-59-7 ADR: 3 III |