Cyclohexylurea 98%

Supplier: Thermo Fisher Scientific

Synonyms: 1-Cyclohexylurea

B24915.14 B24915.22
B24915.14EA 32.7 EUR
B24915.14 B24915.22
Cyclohexylurea 98%
Cyclohexylurea

Purity: 98% CAS No.: 698-90-8 Molecular Formula: C7H14N2O Formula Weight: 142.20 Melting Point: 191-195° Beilstein Reference: 636914 MDL No.: MFCD00014286

Formula: C₇H₁₄N₂O
MW: 142.20 g/mol
Melting Pt: 191…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00014286
CAS Number: 698-90-8
EINECS: 211-822-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR