3-n-Propyl-2-pyrazolin-5-one 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B25214.06
B25214.14
B25214.06EA
53
EUR
B25214.06
B25214.14
3-n-Propyl-2-pyrazolin-5-one 98%
3-n-Propyl-2-pyrazolin-5-one
Purity: 98%
CAS No.: 29211-70-9
Molecular Formula: C6H10N2O
Formula Weight: 126.16
Melting Point: 206-210°
Beilstein Reference: 878994
MDL No.: MFCD00051646
Formula:
C₆H₁₀N₂O Melting Pt: 206…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051646 CAS Number: 29211-70-9 |