1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Supplier: Thermo Fisher Scientific
Warning

Synonyms: DOTA-amide, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetamide

H26792.MD H26792.03
H26792.MDEA 129 EUR
H26792.MD H26792.03
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

CAS No.: 157599-02-5 Molecular Formula: C16H32N8O4 Formula Weight: 400.48 MDL No.: MFCD09263318

Formula: C₁₆H₃₂N₈O₄
MW: 400.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263318
CAS Number: 157599-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR