1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
H27886.03 H27886.06
H27886.03EA 139 EUR
H27886.03 H27886.06
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
CAS No.: 129799-08-2 Molecular Formula: C11H20N2O4 Formula Weight: 244.29 MDL No.: MFCD01632465
Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632465
CAS Number: 129799-08-2