1,1'-Binaphthyl-2,2'-diamine 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: BINAM, [1,1'-Binaphthalene]-2,2'-diamine, racemic-2,2'-Diamino-1,1'-binaphthyl, 2,2'-Diamino-1,1'-binaphthyl, (±)-1,1'-Bi(2-naphthylamine)
H31264.03 H31264.06
H31264.03EA 99.1 EUR
H31264.03 H31264.06
1,1'-Binaphthyl-2,2'-diamine 97%
1,1'-Binaphthyl-2,2'-diamine
Purity: 97% CAS No.: 4488-22-6 Molecular Formula: C20H16N2 Formula Weight: 284.36 Melting Point: 188-193° MDL No.: MFCD00145204
Formula: C₂₀H₁₆N₂
MW: 284.36 g/mol
Melting Pt: 188…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 4488-22-6