3,4-Pyridinedicarboximide
Supplier: Thermo Fisher Scientific
|
Warning
|
H31758.06
H31758.14
H31758.22
H31758.06EA
47.8
EUR
H31758.06
H31758.14
H31758.22
3,4-Pyridinedicarboximide
3,4-Pyridinedicarboximide
CAS No.: 4664-01-1
Molecular Formula: C7H4N2O2
Formula Weight: 148.12
Melting Point: 230-232°
MDL No.: MFCD00013439
Formula:
C₇H₄N₂O₂ Melting Pt: 230…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013439 CAS Number: 4664-01-1 |