Tungsten ≥99.95% (metals basis), rods 2 mm (0.08 in) Ø
Supplier: Thermo Fisher Scientific
43971.BV 43971.9X 43971.CE
43971.BVEA 96.9 EUR
43971.BV 43971.9X 43971.CE
Tungsten ≥99.95% (metals basis), rods 2 mm (0.08 in) Ø
Tungsten
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945