Cyclopropanecarbothioamide 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H32315.03
H32315.06
H32315.03EA
78.8
EUR
H32315.03
H32315.06
Cyclopropanecarbothioamide 97%
Cyclopropanecarbothioamide
Purity: 97%
CAS No.: 20295-34-5
Molecular Formula: C4H7NS
Formula Weight: 101.17
Melting Point: 111-113°
MDL No.: MFCD09469287
Formula:
C₄H₇NS Melting Pt: 111…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD09469287 CAS Number: 20295-34-5 |