2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
Supplier: Thermo Fisher Scientific
Synonyms: Benzylboronic Acid Pinacol Ester
H32523.03 H32523.06 H32523.14
H32523.03EA 51.2 EUR
H32523.03 H32523.06 H32523.14
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity: 96% CAS No.: 87100-28-5 Molecular Formula: C13H19BO2 Formula Weight: 218.1 Boiling Point: 65°/0.15mm MDL No.: MFCD05663841
Formula: C₁₃H₁₉BO₂
Boiling Pt: 65 °C (0.15 mmHg)
Density: 0.98
Storage Temperature: Ambient
MDL Number: MFCD05663841
CAS Number: 87100-28-5