5-Amino-4-(4-fluorophenyl)-1H-pyrazole 97%

Supplier: Thermo Scientific
Warning

H32831.03EA 58.6 EUR
H32831.03
5-Amino-4-(4-fluorophenyl)-1H-pyrazole 97%
5-Amino-4-(4-fluorophenyl)-1H-pyrazole
Formula: C₉H₈FN₃
MW: 177,18 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03001202
CAS Number: 5848-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR