3-(4-Chloro-3-fluorophenyl)propionic acid 96%
Supplier: Thermo Fisher Scientific
Warning
H34205.03 H34205.06
H34205.03EA 40.6 EUR
H34205.03 H34205.06
3-(4-Chloro-3-fluorophenyl)propionic acid 96%
3-(4-Chloro-3-fluorophenyl)propionic acid
Purity: 96% CAS No.: 881189-65-7 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 80-84° MDL No.: MFCD04116058
Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04116058
CAS Number: 881189-65-7