(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-1-BOC-3-(hydroxymethyl)piperidine, (R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(R)-N-BOC-PIPERIDINE METHANOL, 1-Piperidinecarboxylicacid, 3-(hydroxymethyl)-, 1,1-dimethylethyl ester, (3R)-
H52559.03
H52559.06
H52559.03EA
53.2
EUR
H52559.03
H52559.06
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate
Purity: 97%
CAS No.: 140695-85-8
Molecular Formula: C11H21NO3
Formula Weight: 215.29
Melting Point: 91-94°
Optical Rotation: -20° (c=2 in methanol)
MDL No.: MFCD02683202
Formula:
C₁₁H₂₁NO₃ Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683202 CAS Number: 140695-85-8 |