tert-Butyl propargyl ether 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
H53480.06
H53480.14
H53480.22
H53480.06EA
48
EUR
H53480.06
H53480.14
H53480.22
tert-Butyl propargyl ether 98%
tert-Butyl propargyl ether
Purity: 98%
CAS No.: 16314-18-4
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 20°(68°F)
Refractive Index: 1.418
Boiling Point: 119°
UN No.: UN1993
MDL No.: MFCD07368651
Formula:
C₇H₁₂O Boiling Pt: 119 °C Density: 0.835 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07368651 CAS Number: 16314-18-4 ADR: 3,II |