4,5-Diazafluoren-9-one 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H56028.03
H56028.06
H56028.03EA
148
EUR
H56028.03
H56028.06
4,5-Diazafluoren-9-one 98%
4,5-Diazafluoren-9-one
Purity: 98%
CAS No.: 50890-67-0
Molecular Formula: C11H6N2O
Formula Weight: 182.18
Melting Point: 214-217°
MDL No.: MFCD00046892
Formula:
C₁₁H₆N₂O Melting Pt: 214…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046892 CAS Number: 50890-67-0 |