9,9-Di-n-dodecyl-2,7-dibromofluorene 98%
Supplier: Thermo Fisher Scientific
Synonyms:
2,7-Dibromo-9,9-didodecyl-9H-fluorene
H56320.14
H56320.22
H56320.14EA
176
EUR
H56320.14
H56320.22
9,9-Di-n-dodecyl-2,7-dibromofluorene 98%
9,9-Di-n-dodecyl-2,7-dibromofluorene
Purity: 98%
CAS No.: 286438-45-7
Molecular Formula: C37H56Br2
Formula Weight: 660.65
Flash Point: 110°(230°F)
Melting Point: 50-55°
Boiling Point: 180°/0.15mm
MDL No.: MFCD03427215
Formula:
C₃₇H₅₆Br₂ Boiling Pt: 180 °C (0.15 mmHg) Melting Pt: 50…55 °C Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03427215 CAS Number: 286438-45-7 |