Bis[2-(di-t-butylphosphino)ethyl]amine 10% (w/w) in THF
Supplier: Thermo Fisher Scientific
Danger
H60493.03 H60493.06 H60493.14
H60493.03EA 52.8 EUR
H60493.03 H60493.06 H60493.14
Bis[2-(di-t-butylphosphino)ethyl]amine 10% (w/w) in THF
Bis[2-(di-t-butylphosphino)ethyl]amine
  • Refractive Index: 1.4180
  • Odour: Unpleasant
CAS No.: 944710-34-3 Molecular Formula: C20H45NP2 Formula Weight: 361.53 Refractive Index: 1.4180 Odour: Unpleasant UN No.: UN1993 MDL No.: MFCD17014018
Formula: C₂₀H₄₅NP₂
MW: 361.535 g/mol
Density: 0.911
Storage Temperature: Ambient
MDL Number: MFCD17014018
CAS Number: 944710-34-3
UN: 1993
ADR: 3 II