(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
N-Boc-3-(2-pyridyl)-L-alanine, Boc-2-Pal-OH
H63037.03
H63037.06
H63037.03EA
114
EUR
H63037.03
H63037.06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
Purity: 98%
CAS No.: 71239-85-5
Molecular Formula: C13H18N2O4
Formula Weight: 266.29
Physical Form: Solid
Beilstein Reference: 6416850
Optical Rotation: -16°(c=1 in methanol)
MDL No.: MFCD00191190
Formula:
C₁₃H₁₈N₂O₄ MW: 266.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00191190 CAS Number: 71239-85-5 |