2-Oxa-6-azaspiro[3.3]heptane oxalate 97%
Supplier: Thermo Fisher Scientific
Danger
H63578.MD H63578.03 H63578.06
H63578.MDEA 80.1 EUR
H63578.MD H63578.03 H63578.06
2-Oxa-6-azaspiro[3.3]heptane oxalate 97%
2-Oxa-6-azaspiro[3.3]heptane oxalate
Purity: 97% CAS No.: 1159599-99-1 Molecular Formula: C5H9NO.C2H2O4 Formula Weight: 189.17 MDL No.: MFCD11976178
Formula: C₇H₁₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD11976178
CAS Number: 1159599-99-1