trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 87583-89-9
Molecular Formula: C8H18N2
Formula Weight: 142.25
Storage Temperature: 2°C to 8°C
Flash Point: 74°(165°F)
Physical Form: Solid
Density: 0.902
Refractive Index: 1.4720
Appearance: Pale brown
Melting Point: 39-44°
Beilstein Reference: 4304824
Optical Rotation: +14° (c=4 in chloroform)
UN No.: UN3259
MDL No.: MFCD00671528
Formula:
C₈H₁₈N₂ Melting Pt: 39…44 °C Density: 0.902 Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00671528 CAS Number: 87583-89-9 ADR: 8,III |