2-Bromoisophthalaldehyde 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H64423.03
H64423.06
H64423.03EA
41.6
EUR
H64423.03
H64423.06
2-Bromoisophthalaldehyde 97%
2-Bromoisophthalaldehyde
Purity: 97%
CAS No.: 79839-49-9
Molecular Formula: C8H5BrO2
Formula Weight: 213.03
Physical Form: Solid
Melting Point: 139-144°
Beilstein Reference: 4740331
MDL No.: MFCD08461034
Formula:
C₈H₅BrO₂ Melting Pt: 139…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD08461034 CAS Number: 79839-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...