Agarose, high melting , medium resolution for DNA separations
Supplier: Thermo Fisher Scientific
J61123.14
J61123.22
J61123.36
J61123.14EA
68.1
EUR
J61123.14
J61123.22
J61123.36
Agarose, high melting , medium resolution for DNA separations
Agarose
CAS No.: 9012-36-6
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00081294
Applications: For standard DNA applications
Storage Temperature:
Ambient |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 EINECS: 232-731-8 |