Polyethylene glycol 3,350 solution 50%
Supplier: Thermo Fisher Scientific
Synonyms: PEG 3,350, PEG 3350
J61495.AE J61495.AK
J61495.AEEA 67.6 EUR
J61495.AE J61495.AK
Polyethylene glycol 3,350 solution 50%
Polyethylene glycol 3,350
CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Physical Form: Viscous liquid Appearance: Colourless MDL No.: MFCD00081839
Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3