Lecithin (from soy bean) ≥90%
Supplier: Thermo Fisher Scientific
J61675.18 J61675.22 J61675.30
J61675.18EA 34.8 EUR
J61675.18 J61675.22 J61675.30
Lecithin (from soy bean) ≥90%
Lecithin
Applications: Naturally occuring lipid that protects cells from oxidation. Useful as an emulsifier
  • Pale brown to yellow Solid
  • Odourless
  • Beilstein Reference: 5209585
Storage Temperature: Freezer
MDL Number: MFCD00147406
CAS Number: 8002-43-5
Merck Index: 12,05452