Kinase buffer I (5X)
Supplier: Thermo Fisher Scientific
J61967.AE
J61967.AK
J61967.AEEA
50.8
EUR
J61967.AE
J61967.AK
Kinase buffer I (5X)
Buffers
Assay Buffers
Physical Form: Liquid
Appearance: Colourless