DNA marker, broad range
Supplier: Thermo Fisher Scientific
J62291.XF
J62291.S0
J62291.XFEA
77
EUR
J62291.XF
J62291.S0
DNA marker, broad range
Electrophoresis Reagents
Molecular Weight Markers
From 250 to 5000 base pairs: 5 bands from 125 to 1000 bp; four bands from 1500 to 5000 bp. Recommended load volume 20 µl.
Physical Form: Liquid