Irinotecan hydrochloride trihydrate 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 100286-90-6
Molecular Formula: C33H38N4O6.HCl.3H2O
Formula Weight: 677.19 (623.15anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: Pale yellow
Melting Point: 256°
Solubility: Soluble in DMSO at 100mg/ml. Soluble in water at 25mg/ml with warming
MDL No.: MFCD01862255
Applications: Topoisomerase I inhibitor
Formula:
C₃₃H₃₈N₄O₆·HCl·3H₂O Melting Pt: 256 °C |
MDL Number:
MFCD01862255 CAS Number: 136572-09-3 Merck Index: 14,5091 |