Ethisterone 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 434-03-7
Molecular Formula: C21H28O2
Formula Weight: 312.45
Physical Form: Powder
Appearance: White to off-white
Melting Point: 263-269°
Beilstein Reference: 1889895
MDL No.: MFCD00003656
Applications: A progestogen that counteracts the estrogenic proliferative effect on the endometrium
Formula:
C₂₁H₂₈O₂ Melting Pt: 263…269 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003656 CAS Number: 434-03-7 EINECS: 207-096-5 Merck Index: 14,3741 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...