L-Cysteine methyl ester hydrochloride

Supplier: Thermo Fisher Scientific
Warning

J64436.14 J64436.22 J64436.30
J64436.14EA 59.6 EUR
J64436.14 J64436.22 J64436.30
L-Cysteine methyl ester hydrochloride
L-Cysteine methyl ester hydrochloride

CAS No.: 18598-63-5 Molecular Formula: C4H9NO2S.HCl Formula Weight: 171.65 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White Melting Point: 142° dec. Solubility: Soluble in water Beilstein Reference: 3685824 MDL No.: MFCD00038998

Formula: C₄H₉NO₂S·HCl
Melting Pt: 142 °C (dec.)
MDL Number: MFCD00038998
CAS Number: 18598-63-5
EINECS: 242-435-0
Merck Index: 14,5785

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR