Gramicidin 95%

Supplier: Thermo Fisher Scientific

J66771.MC J66771.03
J66771.MCEA 37.3 EUR
J66771.MC J66771.03
Gramicidin 95%
Gramicidin

Purity: 95% CAS No.: 1405-97-6 Molecular Formula: C99H140N20O17 Formula Weight: 1882.29 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 229-230° Solubility: Soluble in acetic acid and lower alcohols. Practically insoluble in water MDL No.: MFCD00131232


Applications: Targets primarily the cell membrane of gram positive bacteria and shows little effect against gram negative bacteria

Formula: C₉₉H₁₄₀N₂₀O₁₇
Melting Pt: 229…230 °C
MDL Number: MFCD00131232
CAS Number: 1405-97-6
EINECS: 215-790-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR