2-Fluorophenylurea 98%
Supplier: Thermo Fisher Scientific
L01247.06 L01247.14
L01247.06EA 28.3 EUR
L01247.06 L01247.14
2-Fluorophenylurea 98%
2-Fluorophenylurea
Purity: 98% CAS No.: 656-31-5 Molecular Formula: C7H7FN2O Formula Weight: 154.14 Melting Point: 178-184° Beilstein Reference: 2803651 MDL No.: MFCD00014786
Formula: C₇H₇FN₂O
MW: 154.14 g/mol
Melting Pt: 178…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00014786
CAS Number: 656-31-5
EINECS: 211-513-6