Dicyclopentadiene diepoxide 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Octahydro-1aH-2,4-methanoindeno[1,2-b:5,6-b']bis(oxirene)
L02940.06
L02940.14
L02940.06EA
58.5
EUR
L02940.06
L02940.14
Dicyclopentadiene diepoxide 98%
Dicyclopentadiene diepoxide
Purity: 99%
CAS No.: 81-21-0
Molecular Formula: C10H12O2
Formula Weight: 164.20
Odour: Odourless
Melting Point: 185-189°
Boiling Point: 120°/10mm subl.
Beilstein Reference: 112743
UN No.: UN2811
MDL No.: MFCD00077209
Formula:
C₁₀H₁₂O₂ MW: 164.20 g/mol Boiling Pt: 326 °C Melting Pt: 185…189 °C Density: 1.331 Storage Temperature: Ambient |
MDL Number:
MFCD00077209 CAS Number: 81-21-0 EINECS: 201-334-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...