1-Cyclopentylethanol 97%
Supplier: Thermo Fisher Scientific
L03446.04
L03446.09
L03446.04EA
91
EUR
L03446.04
L03446.09
1-Cyclopentylethanol 97%
1-Cyclopentylethanol
Purity: 97%
CAS No.: 52829-98-8
Molecular Formula: C7H14O
Formula Weight: 114.19
Refractive Index: 1.4580
Boiling Point: 68°/8mm
Beilstein Reference: 2069851
MDL No.: MFCD00013743
Formula:
C₇H₁₄O Boiling Pt: 68 °C (8 mmHg) Density: 0.911 Storage Temperature: Ambient |
MDL Number:
MFCD00013743 CAS Number: 52829-98-8 EINECS: 258-208-4 |