Triethylsilanol 97%
Supplier: Thermo Fisher Scientific
Synonyms:
Hydroxytriethylsilane, Triethyl(hydroxy)silane
L03738.06
L03738.03
L03738.06EA
106
EUR
L03738.06
L03738.03
Triethylsilanol 97%
Triethylsilanol
Purity: 97%
CAS No.: 597-52-4
Molecular Formula: C6H16OSi
Formula Weight: 132.28
Storage Temperature: 2°C to 8°C
Flash Point: 57°(135°F)
Refractive Index: 1.4330
Odour: Mild
Boiling Point: 154°
Beilstein Reference: 1732848
UN No.: UN1993
MDL No.: MFCD00042640
Formula:
C₆H₁₆OSi MW: 132.28 g/mol |
MDL Number:
MFCD00042640 CAS Number: 597-52-4 |