6-Heptenenitrile 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
L13137.03
L13137.06
L13137.03EA
72.3
EUR
L13137.03
L13137.06
6-Heptenenitrile 96%
6-Heptenenitrile
Purity: 97%
CAS No.: 5048-25-9
Molecular Formula: C7H11N
Formula Weight: 109.17
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₁₁N Boiling Pt: 75…78 °C (13 mmHg) Flash Pt: >100 °C (212 °F) |
MDL Number:
MFCD00151804 CAS Number: 5048-25-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...